Imprimer

Anti-VIM Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F310]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIV2122-03CEA 679 EUR
USBIV2122-03C
Anti-VIM Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F310]
Anticorps
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: VIM (vimentin)
Clonality: monoclonal
Clone: 5F310
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Chicken, Dog, Human, Monkey, Xenopus laevis
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR