Imprimer
behrotest® nitrogen station, KOSTA 2
Fournisseur: Behr
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BEHRB00217639EA 518 EUR
BEHRB00217639
behrotest® nitrogen station, KOSTA 2
Systèmes de digestion Systèmes de minéralisation
In the behrotest® nitrogen station, solvent is evaporated under ambient temperature conditions by using a mild nitrogen flow. A distributor leads the nitrogen flow through two individually adjustable tubes into the Kuderna-Danish flasks. Thus the two samples reduce simulataneously.
The organic phase can be reduced down to less than 1 ml under a mild nitrogen flow.
Certifications: ISO 9377-2