Imprimer
Freezing medium for primary cells, Cryo-SFM
Fournisseur: PromoCell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-29910EA 65 EUR
C-29910 C-29912
Freezing medium for primary cells, Cryo-SFM
Milieux de cryoconservation cellulaire
PromoCell Cryo-SFM is a chemically defined, serum-free and animal component-free cryopreservation medium with DMSO, methylcellulose and other cryoprotective components developed for the cryopreservation of human and animal primary cells. It ensures high cell viability, excellent growth rates and good attachment of the cells after thawing.