Imprimer
Human saphenous vein endothelial cells
Fournisseur: PromoCell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C-14025EA 302 EUR
C-14025 C-12231
Human saphenous vein endothelial cells
Cellules
Primary Human Saphenous Vein Endothelial Cells (HSaVEC) are isolated from human saphe-nous veins from single donors
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
The saphenous vein is often used for autotransplantation in coronary artery bypass operations, when arterial grafts are not available or many grafts are required.
Informations de commande: Human Saphenous Vein Endothelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® area also available.