Imprimer
Grounding cables
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MSGA19331EA 94.2 EUR
MSGA19331 MSGA19310 MSGA19321 MSGA19313 MSGA19323 MSGA19312 MSGA19326 MSGA19325 MSGA2794 MSGA19317 MSGA19327 MSGA19319 MSGA19329
Grounding cables
These earthing cables prevent the danger of explosions as a result of arcing and electrostatic charge.
  • Available in two lengths and either galvanised or double-insulated
  • Can be used for a variety of applications thanks to a combination of various connection options (crocodile clips, screwable C clips, earthing pliers, ring cable lug)