Imprimer
Water purification system, Elix® Essential
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Elix® Essential
MILLZLXEV150SZEA 11060 EUR
MILLZLXEV150SZ MILLZLXEV030UK MILLZLXE0150IT MILLZLXE0030SP MILLZLXEV050GE MILLZLXE0030GE MILLZLXEV050SP MILLZLXE0150EU MILLZLXE0030SZ MILLZLXEV100BE MILLZLXEV100EU MILLZLXEV030IT MILLZLXE0030FR MILLZLXEV150SP MILLZLXEV100IT MILLZLXEV150GE MILLZLXE0150BE MILLZLXEV050SZ MILLZLXE0100SZ MILLZLXE0050UK MILLZLXEV150DK MILLZLXEV100GE MILLZLXE0150FR MILLZLXE0100GE MILLZLXEV100SP MILLZLXE0100SP MILLZLXE0030BE MILLZLXEV100SZ MILLZLXEV030EU MILLZLXE0050DK MILLZLXE0030IT MILLZLXEV100FR MILLZLXE0150SP MILLZLXE0150GE MILLZLXE0100FR MILLZLXEV030FR MILLZLXE0030EU MILLZLXEV050DK MILLZLXE0150SZ MILLZLXE0100BE MILLZLXE0050SP MILLZLXEV150IT MILLZLXE0050GE MILLZLXEV030SP MILLZLXEV100DK MILLZLXEV050UK MILLZLXE0050SZ MILLZLXE0050FR MILLZLXEV030SZ MILLZLXEV050IT MILLZLXE0100EU MILLZLXE0100IT MILLZLXEV030GE MILLZLXE0150DK MILLZLXE0030UK MILLZLXEV150UK MILLZLXEV150FR MILLZLXEV050EU MILLZLXE0050BE MILLZLXE0100UK MILLZLXEV150BE MILLZLXE0150UK MILLZLXE0030DK MILLZLXEV030DK MILLZLXE0050IT MILLZLXE0050EU MILLZLXEV050FR MILLZLXEV050BE MILLZLXEV150EU MILLZLXEV100UK MILLZLXE0100DK
Water purification system, Elix® Essential
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
Elix® Essential delivers Type 2 water customised to fit a broad range of research areas. It is designed to be fed from potable water and is the ideal feed for any laboratory equipment, including ultrapure water systems such as Milli-Q® systems.
  • Depending on daily volume needs, different production flow rates are available (3, 5, 10 & 15 l/h)
  • Pure water is stored in a polyethylene tank designed to minimise risks of contamination during water storage
  • Vent filter provides effective protection against airborne contaminants; ASM (Automatic Sanitization Module) prevents biofilm formation
  • Water can be either sourced through the tank front valve or sent by a distribution pump to feed laboratory instruments
  • Progard® cartridge incorporates RFID technology for instant visibility on cartridge parameters and full traceability
  • Colour-coded backlit LCD display clearly shows message importance with system alert and alarm icons
  • Small footprint enables installation wherever it is convenient in the lab — on or under the bench, or on a wall
  • Millitrack® e-Solutions provide enhanced data management control and remote access capabilities to the system dashboard
To obtain Elix® pure water quality, the purification sequence has been fully optimised. The patented Elix® module (EDI technology) is combined with Progard® complete pretreatment, intelligent reverse osmosis and optional bactericidal UV lamp technologies (in Elix® Essential UV systems). Only Elix® systems are equipped with the unique Elix® module. It provides a superior and consistent water quality while preserving the environment (self-regeneration of resin beads, minimal electricity consumption and no extra softener required). Intelligent Reverse Osmosis controls water consumption, ensures a constant product flow rate and optimal water quality. All prolong the lifetime of the RO membrane and downstream purification technologies.
A single replaceable cartridge, an optional long-lasting UV lamp coupled with timely service alerts, allows the system to run smoothly all year long while preserving budget.
Informations de commande: Progard S2 pretreatment pack and vent filter (171-0590) should be ordered with the main unit.