Imprimer
Pressure regulator
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PERC02400085EA 3050 EUR
PERC02400085
Pressure regulator
Régulateurs de gaz
Two-stage regulator for precise outlet pressure control of helium gas. Made of brass body and stainless steel diaphragm for long life and minimum maintenance.
Maximum inlet pressure of 20700 KPa.
Approximate weight: 1,3 kg