Imprimer
Blades for safety knives, No. 142
Fournisseur: Martor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
233-2238EA 18.5 EUR
233-2238
Blades for safety knives, No. 142
Rasoirs et lames
Non corrosive trapezoid blade for use with safety knives.
  • Deep-edged, two-sided ground-edge
  • Double bevel edge-ground
  • Two-times usable blade
Mountable in:
SECUNORM PROFI
SECUNORM IDEAL