Imprimer
Graduated pipettes, BLAUBRAND®, type 3, Class AS, total delivery (one batch)
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BLAUBRAND®
BRND2.27706EA 83.9 EUR
BRND2.27706 BRND2.27715 BRND2.27705 BRND2.27713 BRND2.27714 BRND2.27711 BRND2.27712 BRND2.27710 BRND2.27709
Graduated pipettes, BLAUBRAND®, type 3, Class AS, total delivery (one batch)
Pipettes Pipettes graduées
AR-Glas®.
  • Calibrated to deliver (TD, Ex); type 3: Zero on top
  • Marks and inscriptions in high contrast blue enamel
  • Waiting time 5 seconds
  • All supplied pipettes come from one batch for statistical testing equipment monitoring
  • Supplied with batch certificate
Length: 360 ±10 mm except for 25 and 50 ml pipettes: 450 ±10 mm.
Certifications: DIN EN ISO 835. DE-M marking.
Attention: * With cotton plug end