Imprimer

Anti-TDO2 Mouse Monoclonal Antibody [clone: 2A4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504879EA 749 EUR
ORIGCF504879
Anti-TDO2 Mouse Monoclonal Antibody [clone: 2A4]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TDO2
Clonality: Monoclonal
Clone: 2A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR