Imprimer

Anti-LRRC25 Mouse Monoclonal Antibody [clone: 1D9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504883EA 770 EUR
ORIGCF504883
Anti-LRRC25 Mouse Monoclonal Antibody [clone: 1D9]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LRRC25
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR