Imprimer

Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 5D7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505672EA 749 EUR
ORIGCF505672
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 5D7]
Anticorps
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 5D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR