Imprimer
Anti-VBP1 Mouse Monoclonal Antibody [clone: 2C2]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF504903EA 749 EUR
ORIGCF504903
Anti-VBP1 Mouse Monoclonal Antibody [clone: 2C2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: VBP1
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human