Imprimer

Anti-FTCD Mouse Monoclonal Antibody [clone: 7C5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504943EA 770 EUR
ORIGCF504943
Anti-FTCD Mouse Monoclonal Antibody [clone: 7C5]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FTCD
Clonality: Monoclonal
Clone: 7C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR