Imprimer

Anti-CST2 Mouse Monoclonal Antibody [clone: 2D6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504560EA 770 EUR
ORIGCF504560
Anti-CST2 Mouse Monoclonal Antibody [clone: 2D6]
Anticorps
Recommended Dilutions: Western Blot: 1:200-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: CST2
Clonality: Monoclonal
Clone: 2D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR