Imprimer

Anti-LRRC50 Mouse Monoclonal Antibody [clone: 3A1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504575EA 749 EUR
ORIGCF504575
Anti-LRRC50 Mouse Monoclonal Antibody [clone: 3A1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LRRC50
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR