Imprimer

Anti-RFC2 Mouse Monoclonal Antibody [clone: 4E1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504956EA 770 EUR
ORIGCF504956
Anti-RFC2 Mouse Monoclonal Antibody [clone: 4E1]
Anticorps
Recommended Dilutions: Western Blot: 1:200-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: RFC2
Clonality: Monoclonal
Clone: 4E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR