Imprimer

Anti-RAB21 Mouse Monoclonal Antibody [clone: 6F6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505738EA 770 EUR
ORIGCF505738
Anti-RAB21 Mouse Monoclonal Antibody [clone: 6F6]
Anticorps
Recommended Dilutions: Western Blot: 1:200-4000

Type: Primary
Antigen: RAB21
Clonality: Monoclonal
Clone: 6F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR