Imprimer

Anti-USP16 Mouse Monoclonal Antibody [clone: 1B4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505771EA 749 EUR
ORIGCF505771
Anti-USP16 Mouse Monoclonal Antibody [clone: 1B4]
Anticorps
Recommended Dilutions: Western Blot: 1:200-4000

Type: Primary
Antigen: USP16
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR