Imprimer
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 4B1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505774EA 749 EUR
ORIGCF505774
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 4B1]
Anticorps
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 4B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat