Imprimer

Anti-USP10 Mouse Monoclonal Antibody [clone: 2E1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF503878EA 770 EUR
ORIGCF503878
Anti-USP10 Mouse Monoclonal Antibody [clone: 2E1]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: USP10
Clonality: Monoclonal
Clone: 2E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR