Imprimer

Anti-CRYZL1 Mouse Monoclonal Antibody [clone: 1D9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504608EA 749 EUR
ORIGCF504608
Anti-CRYZL1 Mouse Monoclonal Antibody [clone: 1D9]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CRYZL1
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR