Imprimer

Anti-HSD17B2 Mouse Monoclonal Antibody [clone: 3E9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504616EA 749 EUR
ORIGCF504616
Anti-HSD17B2 Mouse Monoclonal Antibody [clone: 3E9]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HSD17B2
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR