Imprimer

Anti-CPA2 Mouse Monoclonal Antibody [clone: 1A1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504644EA 770 EUR
ORIGCF504644
Anti-CPA2 Mouse Monoclonal Antibody [clone: 1A1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CPA2
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR