Imprimer

Anti-ESRRG Mouse Monoclonal Antibody [clone: 1C2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505026EA 749 EUR
ORIGCF505026
Anti-ESRRG Mouse Monoclonal Antibody [clone: 1C2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ESRRG
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR