Imprimer
Anti-ZBED1 Mouse Monoclonal Antibody [clone: 2E5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505042EA 749 EUR
ORIGCF505042
Anti-ZBED1 Mouse Monoclonal Antibody [clone: 2E5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: ZBED1
Clonality: Monoclonal
Clone: 2E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human