Imprimer

Anti-SYT4 Mouse Monoclonal Antibody [clone: 3B5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505864EA 770 EUR
ORIGCF505864
Anti-SYT4 Mouse Monoclonal Antibody [clone: 3B5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SYT4
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR