Imprimer

Anti-MAPK13 Mouse Monoclonal Antibody [clone: 2G6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505865EA 749 EUR
ORIGCF505865
Anti-MAPK13 Mouse Monoclonal Antibody [clone: 2G6]
Anticorps
Recommended Dilutions: Western Blot: 1:200-1000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK13
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR