Imprimer
Anti-MAPK9 Mouse Monoclonal Antibody [clone: 2D11]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505877EA 764 EUR
ORIGCF505877
Anti-MAPK9 Mouse Monoclonal Antibody [clone: 2D11]
Anticorps
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150
Type: Primary
Antigen: MAPK9
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat