Imprimer

Anti-PDIA4 Mouse Monoclonal Antibody [clone: 2A11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF503889EA 764 EUR
ORIGCF503889
Anti-PDIA4 Mouse Monoclonal Antibody [clone: 2A11]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: PDIA4
Clonality: Monoclonal
Clone: 2A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR