Imprimer
Anti-PDIA4 Mouse Monoclonal Antibody [clone: 6F10]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503887EA 749 EUR
ORIGCF503887
Anti-PDIA4 Mouse Monoclonal Antibody [clone: 6F10]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: PDIA4
Clonality: Monoclonal
Clone: 6F10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human