Imprimer
Anti-RYBP Mouse Monoclonal Antibody [clone: 2D4]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF504652EA 749 EUR
ORIGCF504652
Anti-RYBP Mouse Monoclonal Antibody [clone: 2D4]
Anticorps
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: RYBP
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human