Imprimer

Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 1C3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504681EA 749 EUR
ORIGCF504681
Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 1C3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: UBE2E3
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR