Imprimer
Anti-ZNF365 Mouse Monoclonal Antibody [clone: 9H5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505065EA 749 EUR
ORIGCF505065
Anti-ZNF365 Mouse Monoclonal Antibody [clone: 9H5]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150
Type: Primary
Antigen: ZNF365
Clonality: Monoclonal
Clone: 9H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human