Imprimer
Anti-TBCC Mouse Monoclonal Antibody [clone: 3A10]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF504734EA 770 EUR
ORIGCF504734
Anti-TBCC Mouse Monoclonal Antibody [clone: 3A10]
Anticorps
Recommended Dilutions: Western Blot: 1:2000-4000
Type: Primary
Antigen: TBCC
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse, Rat