Imprimer

Anti-NDUFB10 Mouse Monoclonal Antibody [clone: 2B4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505114EA 770 EUR
ORIGCF505114
Anti-NDUFB10 Mouse Monoclonal Antibody [clone: 2B4]
Anticorps
Recommended Dilutions: Western Blot: 1:200-1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NDUFB10
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR