Imprimer
Anti-FAM164A Mouse Monoclonal Antibody [clone: 2D1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505122EA 749 EUR
ORIGCF505122
Anti-FAM164A Mouse Monoclonal Antibody [clone: 2D1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: FAM164A
Clonality: Monoclonal
Clone: 2D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human