Imprimer

Anti-BBOX1 Mouse Monoclonal Antibody [clone: 8B12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505786EA 749 EUR
ORIGCF505786
Anti-BBOX1 Mouse Monoclonal Antibody [clone: 8B12]
Anticorps
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: BBOX1
Clonality: Monoclonal
Clone: 8B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR