Imprimer

Anti-ERBB2 Mouse Monoclonal Antibody [clone: 6F3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505802EA 749 EUR
ORIGCF505802
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 6F3]
Anticorps
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 6F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR