Imprimer
Anti-USP10 Mouse Monoclonal Antibody [clone: 1H9]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF504007EA 770 EUR
ORIGCF504007
Anti-USP10 Mouse Monoclonal Antibody [clone: 1H9]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: USP10
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey