Imprimer

Anti-PECAM1 Mouse Monoclonal Antibody [clone: 1B10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504763EA 749 EUR
ORIGCF504763
Anti-PECAM1 Mouse Monoclonal Antibody [clone: 1B10]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR