Imprimer
Anti-UBE2S Mouse Monoclonal Antibody [clone: 4B8]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505169EA 749 EUR
ORIGCF505169
Anti-UBE2S Mouse Monoclonal Antibody [clone: 4B8]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: UBE2S
Clonality: Monoclonal
Clone: 4B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human