Imprimer
Anti-RALB Mouse Monoclonal Antibody [clone: 2C4]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505880EA 749 EUR
ORIGCF505880
Anti-RALB Mouse Monoclonal Antibody [clone: 2C4]
Anticorps
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: RALB
Clonality: Monoclonal
Clone: 2C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat