Imprimer
Anti-CALR3 Mouse Monoclonal Antibody [clone: 1E1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505886EA 749 EUR
ORIGCF505886
Anti-CALR3 Mouse Monoclonal Antibody [clone: 1E1]
Anticorps
Recommended Dilutions: Western Blot: 1:200-2000
Type: Primary
Antigen: CALR3
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat