Imprimer
Anti-RASA1 Mouse Monoclonal Antibody [clone: 1A4]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505901EA 749 EUR
ORIGCF505901
Anti-RASA1 Mouse Monoclonal Antibody [clone: 1A4]
Anticorps
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: RASA1
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human