Imprimer

Anti-NONO Mouse Monoclonal Antibody [clone: 5F1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504802EA 749 EUR
ORIGCF504802
Anti-NONO Mouse Monoclonal Antibody [clone: 5F1]
Anticorps
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150

Type: Primary
Antigen: NONO
Clonality: Monoclonal
Clone: 5F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR