Imprimer
Anti-PADI4 Mouse Monoclonal Antibody [clone: 5E7]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF504814EA 749 EUR
ORIGCF504814
Anti-PADI4 Mouse Monoclonal Antibody [clone: 5E7]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150
Type: Primary
Antigen: PADI4
Clonality: Monoclonal
Clone: 5E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human