Imprimer
Anti-C2orf43 Mouse Monoclonal Antibody [clone: 2G9]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF504833EA 749 EUR
ORIGCF504833
Anti-C2orf43 Mouse Monoclonal Antibody [clone: 2G9]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: C2orf43
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human