Imprimer
Anti-ESRRG Mouse Monoclonal Antibody [clone: 6G1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505197EA 749 EUR
ORIGCF505197
Anti-ESRRG Mouse Monoclonal Antibody [clone: 6G1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: ESRRG
Clonality: Monoclonal
Clone: 6G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human