Imprimer
Anti-PIK3CG Mouse Monoclonal Antibody [clone: 6H5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505228EA 749 EUR
ORIGCF505228
Anti-PIK3CG Mouse Monoclonal Antibody [clone: 6H5]
Anticorps
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: PIK3CG
Clonality: Monoclonal
Clone: 6H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse